Assay Details
| Gene Symbol: Tal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287388.1 | NP_001274317.1 | 2 - 3 | 211 | 96 | |
| NM_011527.3 | NP_035657.1 | 2 - 3 | 194 | 96 | ||
| GenBank mRNA | AK144164.1 | - | 1 - 2 | 164 | 96 | |
| AK157852.1 | - | 2 - 3 | 212 | 96 | ||
| BC063060.1 | - | 2 - 3 | 194 | 6826611 | 96 | |
| M59764.1 | - | 1 - 2 | 142 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK157852.1, BC063060.1, M59764.1, NM_001287388.1, NM_011527.3 transcripts.
Set Membership: |
|

Tal1
Genomic Map