Assay Details
| Gene Symbol: Stil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009185.3 | NP_033211.2 | 12 - 13 | 2460 | 70 | |
| GenBank mRNA | AK019471.1 | - | 1 - 2 | 813 | 70 | |
| AK049223.1 | - | 12 - 13 | 2397 | 70 | ||
| BC049865.1 | - | 12 - 13 | 2414 | 70 | ||
| BC060706.1 | - | 13 - 14 | 2482 | 70 | ||
| BC108373.1 | - | 11 - 12 | 2128 | 70 | ||
| BC141175.1 | - | 12 - 13 | 2333 | 9055787 | 70 | |
| BC145493.1 | - | 12 - 13 | 2333 | 9054056 | 70 | |
| U36778.1 | - | 12 - 13 | 2378 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stil
Genomic Map