Assay Details
| Gene Symbol: Foxd2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078937.1 | - | 407 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box D2 | Foxd2 |
AI426778, Mf2 |
-359.0 | Anti-sense | Chr. 4 - 114906280 - 114908898 on NCBI Build GRCm38 |
| Downstream | forkhead box E3 | Foxe3 |
FREAC8, dyl, rct |
8.8kb | Anti-sense | Chr. 4 - 114925147 - 114926013 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Foxd2os
Genomic Map