Assay Details
| Gene Symbol: Eri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285899.1 | NP_001272828.1 | 5 - 6 | 911 | 80 | |
| NM_001285901.1 | NP_001272830.1 | 4 - 5 | 494 | 80 | ||
| NM_001285902.1 | NP_001272831.1 | 3 - 4 | 216 | 80 | ||
| NM_080469.4 | NP_536717.2 | 5 - 6 | 897 | 80 | ||
| GenBank mRNA | AA259566.1 | - | 3 - 4 | 206 | 80 | |
| AK083017.1 | - | 5 - 6 | 768 | 80 | ||
| AK144107.1 | - | 5 - 6 | 899 | 80 | ||
| AY029599.1 | - | 2 - 3 | 90 | 80 | ||
| BC021405.1 | - | 3 - 4 | 447 | 80 | ||
| BC079640.1 | - | 1 - 2 | 61 | 80 | ||
| BC096545.1 | - | 5 - 6 | 3044 | 80 | ||
| BC116685.1 | - | 5 - 6 | 744 | 40105869 | 80 | |
| BC117540.1 | - | 5 - 6 | 758 | 40105872 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eri3
Genomic Map