Assay Details
| Gene Symbol: Eif2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111277.1 | NP_001104747.1 | 1 - 2 | 62 | 64 | |
| NM_175135.4 | NP_780344.2 | 1 - 2 | 62 | 64 | ||
| GenBank mRNA | AK145856.1 | - | 1 - 2 | 39 | 64 | |
| AK172173.1 | - | 1 - 2 | 56 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145856.1, NM_001111277.1, NM_175135.4 transcripts.
Set Membership: |
|

Eif2b3
Genomic Map