Assay Details
| Gene Symbol: Eif2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111277.1 | NP_001104747.1 | 5 - 6 | 646 | 85 | |
| NM_175135.4 | NP_780344.2 | 5 - 6 | 646 | 85 | ||
| GenBank mRNA | AK145856.1 | - | 5 - 6 | 623 | 85 | |
| AK172302.1 | - | 5 - 6 | 596 | 85 | ||
| BC115657.1 | - | 3 - 4 | 320 | 40103205 | 85 | |
| BC115658.1 | - | 3 - 4 | 320 | 40103206 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif2b3
Genomic Map