Assay Details
| Gene Symbol: Gm12992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102393.1 | 1 - 2 | 259 | 76 | ||
| NR_102394.1 | 1 - 2 | 259 | 76 | |||
| GenBank mRNA | BE655205.1 | - | 1 - 2 | 259 | 76 | |
More Information
Set Membership: |
|

Gm12992
Genomic Map