Assay Details
| Gene Symbol: Gm12992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102393.1 | 2 - 3 | 338 | 66 | ||
| GenBank mRNA | AK047558.1 | - | 1 - 2 | 76 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gm12992
Genomic Map