Assay Details
| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053069.5 | NP_444299.1 | 2 - 3 | 399 | 113 | |
| GenBank mRNA | AB048349.1 | - | 1 - 2 | 114 | 113 | |
| AK028315.1 | - | 2 - 3 | 401 | 113 | ||
| BC002166.1 | - | 2 - 3 | 265 | 3486093 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg5
Genomic Map