Assay Details
| Gene Symbol: Atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053069.5 | NP_444299.1 | 4 - 5 | 601 | 72 | |
| GenBank mRNA | AB048349.1 | - | 3 - 4 | 316 | 72 | |
| AK028315.1 | - | 4 - 5 | 603 | 72 | ||
| BC002166.1 | - | 4 - 5 | 467 | 3486093 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg5
Genomic Map