Assay Details
| Gene Symbol: Sesn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144907.1 | NP_659156.1 | 5 - 6 | 1050 | 78 | |
| GenBank mRNA | AK143788.1 | - | 5 - 6 | 1094 | 78 | |
| AK170228.1 | - | 5 - 6 | 1063 | 78 | ||
| AK170547.1 | - | 1094 | 78 | |||
| BC005672.1 | - | 5 - 6 | 1050 | 3153129 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sesn2
Genomic Map