Assay Details
| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008698.2 | NP_032724.1 | 9 - 10 | 797 | 90 | |
| GenBank mRNA | AJ001260.1 | - | 9 - 10 | 797 | 90 | |
| AK031804.1 | - | 9 - 10 | 813 | 90 | ||
| AK147786.1 | - | 10 - 11 | 1074 | 90 | ||
| BC010837.1 | - | 9 - 10 | 795 | 4242309 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124016 | NM_008698.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 424479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654643 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658998 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | AB030187.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | AK013203.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | AK018154.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | AK163183.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | BC027164.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 726025 | NM_022424.5 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870846 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198567 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57277 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149012 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110702 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124016 | NM_008698.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214516 | AJ001260.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P214516 | NM_008698.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560176 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560172 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27560167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110702 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07241873 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07241858 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149012 | - | - |
| Whole Mouse Genome G4122A | A_51_P214516 | AJ001260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P214516 | NM_008698.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117105 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110702 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09516254 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09516250 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09177186 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09516251 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09516249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27560159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09155180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09177185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09352214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09489194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09721473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09399831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09249260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09226074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09315477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09516249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09584473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09226064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27560164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10373962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373961 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373959 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373956 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373960 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373958 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448465_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456748_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373950 | - | - |
| Mouse Genome 430 2.0 Array | 1448465_at | - | - |
| Mouse Genome 430 2.0 Array | 1448465_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456748_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456748_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18082_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18082_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100000613.mm.1 | NM_008698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000613.mm.1 | NM_008698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000613.mm.1 | TC1100000053.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000615.mm.1 | NM_008698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000615.mm.1 | NM_008698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000615.mm.1 | TC1100000053.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100000614.mm.1 | NM_008698.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000614.mm.1 | NM_008698.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100000614.mm.1 | TC1100000053.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448465_at | - | - |
| Mouse Expression Set 430 | 1448465_PM_at | - | - |
| Mouse Expression Set 430 | 1456748_a_at | - | - |
| Mouse Expression Set 430 | 1456748_PM_a_at | - | - |
| Murine Genome U74v2 Set | 93251_PM_at | - | - |
| Murine Genome U74v2 Set | 93251_at | - | - |
| Murine Genome U74v2 Set | 132609_g_at | - | - |
| Murine Genome U74v2 Set | 132608_PM_at | - | - |
| Murine Genome U74v2 Set | 132608_at | - | - |
| Murine Genome U74v2 Set | 132609_PM_g_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448465_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456748_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456748_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448465_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448465_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456748_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456748_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448465_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 918513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6860138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860138 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619861 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619861 | - | - |
| mouse-6 v1.1 (Illumina) | scl41882.13_532-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860138 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41882.13_532-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860138 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2619861 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619861 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41882.13_532-S | - | - |

Nipsnap1
Genomic Map