Assay Details
| Gene Symbol: Tmed4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134020.1 | NP_598781.1 | 3 - 4 | 385 | 63 | |
| GenBank mRNA | AF368921.1 | - | 3 - 4 | 416 | 63 | |
| AK078425.1 | - | 3 - 4 | 396 | 63 | ||
| AK158876.1 | - | 3 - 4 | 418 | 63 | ||
| AK204643.1 | - | 1 - 2 | 80 | 63 | ||
| BC023041.1 | - | 3 - 4 | 385 | 5360236 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmed4
Genomic Map