Assay Details
| Gene Symbol: Hectd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175244.3 | NP_780453.1 | 19 - 20 | 2488 | 78 | |
| GenBank mRNA | AK051385.1 | - | 19 - 20 | 2501 | 78 | |
| AK139305.1 | - | 19 - 20 | 2516 | 78 | ||
| AK154378.1 | - | 19 - 20 | 2520 | 78 | ||
| AK155290.1 | - | 19 - 20 | 2501 | 78 | ||
| AK155676.1 | - | 19 - 20 | 2501 | 78 | ||
| AK163323.1 | - | 19 - 20 | 2523 | 78 | ||
| AK165482.1 | - | 19 - 20 | 2520 | 78 | ||
| AK170930.1 | - | 19 - 20 | 2501 | 78 | ||
| BC024415.1 | - | 5 - 6 | 556 | 78 | ||
| BC070411.1 | - | 19 - 20 | 2488 | 5706209 | 78 | |
| BC082276.1 | - | 9 - 10 | 942 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hectd3
Genomic Map