| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400027075.mm.1 |
NM_023178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027075.mm.1 |
NM_023178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027075.mm.1 |
TC0400003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027077.mm.1 |
NM_023178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027077.mm.1 |
NM_023178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027077.mm.1 |
TC0400003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027078.mm.1 |
NM_023178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027078.mm.1 |
NM_023178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027078.mm.1 |
TC0400003373.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400027080.mm.1 |
NM_023178.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027080.mm.1 |
NM_023178.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400027080.mm.1 |
TC0400003373.mm.1 |
MTA TC |