Assay Details
| Gene Symbol: Lypla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK003689.1 | - | 1 - 2 | 40 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003689.1 transcripts.
Set Membership: |
|

Lypla2
Genomic Map