Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009213.2 | NP_033239.1 | 2 - 3 | 456 | 91 | |
| GenBank mRNA | AJ222800.1 | - | 2 - 3 | 457 | 91 | |
| AK075616.1 | - | 2 - 3 | 466 | 91 | ||
| AK088962.1 | - | 1 - 2 | 869 | 91 | ||
| AK161573.1 | - | 870 | 91 | |||
| BC010978.1 | - | 2 - 3 | 466 | 4193282 | 91 | |
| BC086487.1 | - | 1 - 2 | 658 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smpd2
Genomic Map