Assay Details
| Gene Symbol: Trim62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178110.2 | NP_835211.1 | 3 - 4 | 798 | 82 | |
| GenBank mRNA | AK048725.1 | - | 5 - 6 | 1637 | 82 | |
| BC049095.1 | - | 3 - 4 | 798 | 6416690 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim62
Genomic Map