Assay Details
| Gene Symbol: Akap12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB070852.1 | - | 1 - 2 | 140 | 97 | |
| AF326230.1 | - | 1 - 2 | 109 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB070852.1, AF326230.1 transcripts.
Set Membership: |
|

Akap12
Genomic Map