Assay Details
| Gene Symbol: Mpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122949.2 | NP_001116421.1 | 9 - 10 | 1505 | 101 | |
| NM_001285496.1 | NP_001272425.1 | 9 - 10 | 1325 | 101 | ||
| NM_010823.3 | NP_034953.2 | 9 - 10 | 1481 | 101 | ||
| GenBank mRNA | AF360122.1 | - | 9 - 10 | 1262 | 101 | |
| AK041166.1 | - | 9 - 10 | 1453 | 101 | ||
| BC103513.1 | - | 9 - 10 | 1438 | 101 | ||
| BC103515.1 | - | 9 - 10 | 1462 | 101 | ||
| BC146612.1 | - | 9 - 10 | 1434 | 101 | ||
| BC146613.1 | - | 9 - 10 | 1439 | 40039147 | 101 | |
| X73677.1 | - | 9 - 10 | 1449 | 101 | ||
| Z22649.1 | - | 9 - 10 | 1439 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpl
Genomic Map