Assay Details
| Gene Symbol: Nhsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163592.1 | NP_001157064.1 | 2 - 3 | 341 | 70 | |
| NM_173390.3 | NP_775566.3 | 2 - 3 | 996 | 70 | ||
| GenBank mRNA | AK017530.1 | - | 2 - 3 | 70 | ||
| AK038892.1 | - | 2 - 3 | 972 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nhsl1
Genomic Map