Assay Details
| Gene Symbol: Mpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122949.2 | NP_001116421.1 | 3 - 4 | 430 | 91 | |
| NM_001285497.1 | NP_001272426.1 | 3 - 4 | 430 | 91 | ||
| NM_010823.3 | NP_034953.2 | 3 - 4 | 430 | 91 | ||
| GenBank mRNA | AK041166.1 | - | 3 - 4 | 402 | 91 | |
| BC101943.1 | - | 3 - 4 | 387 | 91 | ||
| BC103513.1 | - | 3 - 4 | 387 | 91 | ||
| BC103514.1 | - | 3 - 4 | 387 | 91 | ||
| BC103515.1 | - | 3 - 4 | 387 | 91 | ||
| BC146612.1 | - | 3 - 4 | 383 | 91 | ||
| BC146613.1 | - | 3 - 4 | 388 | 40039147 | 91 | |
| X73677.1 | - | 3 - 4 | 374 | 91 | ||
| Z22649.1 | - | 3 - 4 | 388 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpl
Genomic Map