Assay Details
| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153403.2 | NP_700452.2 | 14 - 15 | 1836 | 162 | |
| GenBank mRNA | AB081471.1 | - | 14 - 15 | 1836 | 162 | |
| AK080954.1 | - | 12 - 13 | 1686 | 162 | ||
| BC129914.1 | - | 14 - 15 | 1836 | 40111808 | 162 | |
| BC129915.1 | - | 14 - 15 | 1832 | 162 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago1
Genomic Map