Assay Details
| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153403.2 | NP_700452.2 | 9 - 10 | 1141 | 65 | |
| GenBank mRNA | AK080954.1 | - | 7 - 8 | 991 | 65 | |
| BC129914.1 | - | 9 - 10 | 1141 | 40111808 | 65 | |
| BC129915.1 | - | 9 - 10 | 1137 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago1
Genomic Map