Assay Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026601.3 | NP_080877.3 | 5 - 6 | 666 | 127 | |
| NR_003946.2 | 5 - 6 | 554 | 127 | |||
| GenBank mRNA | AK004328.1 | - | 4 - 5 | 254 | 127 | |
| AK020099.1 | - | 5 - 6 | 540 | 127 | ||
| BC024619.1 | - | 5 - 6 | 654 | 4923664 | 127 | |
| BC100401.1 | - | 5 - 6 | 630 | 127 | ||
| CB952524.1 | - | 3 - 4 | 171 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hyi
Genomic Map