Assay Details
| Gene Symbol: Hyi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026601.3 | NP_080877.3 | 6 - 7 | 791 | 73 | |
| NR_003946.2 | 6 - 7 | 679 | 73 | |||
| GenBank mRNA | AK004328.1 | - | 5 - 6 | 379 | 73 | |
| BC024619.1 | - | 6 - 7 | 779 | 4923664 | 73 | |
| BC100401.1 | - | 6 - 7 | 755 | 73 | ||
| BX525112.1 | - | 4 - 5 | 232 | 73 | ||
| CB952524.1 | - | 4 - 5 | 296 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239810 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239809 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239808 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135039 | BX525112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135039 | NR_003946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135039 | NM_026601.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709943 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126253 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164830 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132427 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135039 | BX525112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135039 | NR_003946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135039 | NM_026601.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239777 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239797 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23239786 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164830 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132427 | - | - |
| Whole Mouse Genome G4122A | A_51_P377579 | BC024619.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P377579 | BC100401.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132427 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164830 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03431454 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03431465 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03431467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03431471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03431460 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239807 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239810 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239806 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239808 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23239809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23239819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03520926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03327518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03352747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03208508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03745292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03859266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239832 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500380 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03681510 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03500368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03654121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03715818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23239758 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10507522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507526 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507528 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10507527 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425201_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10507520 | - | - |
| Mouse Genome 430 2.0 Array | 1425201_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425201_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68180_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68180_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400009795.mm.1 | NR_003946.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009795.mm.1 | NR_003946.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400009795.mm.1 | TC0400001305.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425201_a_at | - | - |
| Mouse Expression Set 430 | 1425201_PM_a_at | - | - |
| Murine Genome U74v2 Set | 169116_at | - | - |
| Murine Genome U74v2 Set | 169116_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425201_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425201_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425201_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425201_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2691567 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2691567 | - | - |

Hyi
Genomic Map