Assay Details
| Gene Symbol: Smyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160127.1 | NP_001153599.1 | 5 - 6 | 782 | 86 | |
| GenBank mRNA | AK142252.1 | - | 5 - 6 | 760 | 86 | |
| BC076601.1 | - | 5 - 6 | 760 | 30609878 | 86 | |
| U76371.1 | - | 887 | 86 | |||
| U76373.2 | - | 5 - 6 | 754 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smyd1
Genomic Map