Assay Details
| Gene Symbol: Wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271757.1 | NP_001258686.1 | 1 - 2 | 170 | 73 | |
| GenBank mRNA | AK049558.1 | - | 1 - 2 | 137 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271757.1 transcripts.
Set Membership: |
|

Wnt5b
Genomic Map