| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600023790.mm.1 |
NM_001134391.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023790.mm.1 |
NM_001134391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023790.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023790.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023790.mm.1 |
TC0600003071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023797.mm.1 |
NM_001134391.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023797.mm.1 |
NM_001134391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023797.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023797.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023797.mm.1 |
TC0600003071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023789.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023789.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023789.mm.1 |
TC0600003071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023793.mm.1 |
NM_001134391.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023793.mm.1 |
NM_001134391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023793.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023793.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023793.mm.1 |
TC0600003071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023788.mm.1 |
NM_001134391.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023788.mm.1 |
NM_001134391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023788.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023788.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023788.mm.1 |
TC0600003071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600023791.mm.1 |
NM_001134391.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023791.mm.1 |
NM_001134391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023791.mm.1 |
NM_031159.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023791.mm.1 |
NM_031159.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600023791.mm.1 |
TC0600003071.mm.1 |
MTA TC |