Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 14 - 15 | 1554 | 126 | |
| GenBank mRNA | AK144547.1 | - | 14 - 15 | 1555 | 126 | |
| AK165019.1 | - | 13 - 14 | 2174 | 126 | ||
| BC059244.1 | - | 5 - 6 | 465 | 126 | ||
| BC144781.1 | - | 14 - 15 | 1497 | 9053329 | 126 | |
| BC145693.1 | - | 14 - 15 | 1497 | 40130996 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map