Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 16 - 17 | 1773 | 138 | |
| GenBank mRNA | AK144547.1 | - | 16 - 17 | 1774 | 138 | |
| AK165019.1 | - | 15 - 16 | 2393 | 138 | ||
| BC059244.1 | - | 7 - 8 | 684 | 138 | ||
| BC144781.1 | - | 16 - 17 | 1716 | 9053329 | 138 | |
| BC145693.1 | - | 16 - 17 | 1716 | 40130996 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map