Assay Details
| Gene Symbol: Exoc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177077.2 | NP_796051.2 | 17 - 18 | 1903 | 111 | |
| GenBank mRNA | AK144547.1 | - | 17 - 18 | 1904 | 111 | |
| AK165019.1 | - | 16 - 17 | 2523 | 111 | ||
| AK220360.1 | - | 2 - 3 | 177 | 111 | ||
| BC059244.1 | - | 8 - 9 | 814 | 111 | ||
| BC144781.1 | - | 17 - 18 | 1846 | 9053329 | 111 | |
| BC145693.1 | - | 17 - 18 | 1846 | 40130996 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc6b
Genomic Map