Assay Details
| Gene Symbol: Ctnna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109764.1 | NP_001103234.1 | 3 - 4 | 462 | 65 | |
| NM_009819.2 | NP_033949.2 | 2 - 3 | 360 | 65 | ||
| NM_145732.2 | NP_663785.2 | 3 - 4 | 462 | 65 | ||
| GenBank mRNA | AK035033.1 | - | 2 - 3 | 256 | 65 | |
| AK158916.1 | - | 3 - 4 | 495 | 65 | ||
| BC051995.1 | - | 2 - 3 | 243 | 65 | ||
| BC079648.1 | - | 3 - 4 | 379 | 6850068 | 65 | |
| CD578966.1 | - | 2 - 3 | 360 | 65 | ||
| D25281.1 | - | 3 - 4 | 389 | 65 | ||
| D25282.1 | - | 2 - 3 | 289 | 65 | ||
More Information
Set Membership: |
|

Ctnna2
Genomic Map