Assay Details
| Gene Symbol: Il17re | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034029.1 | NP_001029201.1 | 2 - 3 | 222 | 110 | |
| NM_001034031.1 | NP_001029203.1 | 4 - 5 | 297 | 110 | ||
| NM_145826.5 | NP_665825.2 | 3 - 4 | 649 | 110 | ||
| GenBank mRNA | AF458068.1 | - | 3 - 4 | 299 | 110 | |
| AK046302.1 | - | 3 - 4 | 652 | 110 | ||
| AK046713.1 | - | 3 - 4 | 311 | 110 | ||
| AK169498.1 | - | 3 - 4 | 477 | 110 | ||
| BC069861.1 | - | 3 - 4 | 326 | 6311568 | 110 | |
| DQ092339.1 | - | 3 - 4 | 238 | 110 | ||
| DQ092340.1 | - | 2 - 3 | 222 | 110 | ||
| DQ092341.1 | - | 4 - 5 | 297 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the DQ092340.1, DQ092341.1, NM_001034029.1, NM_001034031.1 transcripts.
Set Membership: |
|

Il17re
Genomic Map