Assay Details
| Gene Symbol: Ppil6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028430.1 | NP_082706.1 | 7 - 8 | 870 | 104 | |
| GenBank mRNA | AK013818.1 | - | 7 - 8 | 832 | 104 | |
| BC147756.1 | - | 7 - 8 | 866 | 9056705 | 104 | |
| BC147757.1 | - | 7 - 8 | 866 | 9056706 | 104 | |
| BP772786.1 | - | 1 - 2 | 104 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942339 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 925143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683295 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853531 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957647 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540282 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491081 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762102 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81658 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176514 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135441 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | NM_028430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | AI463820.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | AK013818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | BC147756.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P224042 | BC147757.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27089707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089682 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08620153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08427823 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08863999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08523387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08620146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089568 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08947534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08354097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27089563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08286500 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10362798 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362797 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362800 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362799 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362796 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362802 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10362801 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453310_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10362794 | - | - |
| Mouse Genome 430 2.0 Array | 1453310_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453310_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73075_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73075_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000002530.mm.1 | NM_028430.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000002530.mm.1 | NM_028430.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000002530.mm.1 | TC1000000366.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000002531.mm.1 | NM_028430.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000002531.mm.1 | NM_028430.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000002531.mm.1 | TC1000000366.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453310_at | - | - |
| Mouse Expression Set 430 | 1453310_PM_at | - | - |
| Murine Genome U74v2 Set | 169115_i_at | - | - |
| Murine Genome U74v2 Set | 166791_PM_at | - | - |
| Murine Genome U74v2 Set | 169115_PM_i_at | - | - |
| Murine Genome U74v2 Set | 166791_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453310_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453310_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2481541 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2481541 | - | - |

Ppil6
Genomic Map