Assay Details
| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050292.1 | - | 2 - 3 | 258 | 97 | |
| BC112401.1 | - | 13 - 14 | 1875 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc162| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050292.1 | - | 2 - 3 | 258 | 97 | |
| BC112401.1 | - | 13 - 14 | 1875 | 97 | ||
Set Membership: |
|