Assay Details
| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050292.1 | - | 4 - 5 | 502 | 89 | |
| BC112401.1 | - | 15 - 16 | 2119 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc162| Gene Symbol: Ccdc162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050292.1 | - | 4 - 5 | 502 | 89 | |
| BC112401.1 | - | 15 - 16 | 2119 | 89 | ||
Set Membership: |
|