Assay Details
| Gene Symbol: Smpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009213.2 | NP_033239.1 | 3 - 4 | 534 | 80 | |
| GenBank mRNA | AJ222800.1 | - | 3 - 4 | 535 | 80 | |
| AK075616.1 | - | 3 - 4 | 544 | 80 | ||
| BC010978.1 | - | 3 - 4 | 544 | 4193282 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smpd2
Genomic Map