Assay Details
| Gene Symbol: Cdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027879.2 | NP_082155.1 | 11 - 12 | 1273 | 85 | |
| GenBank mRNA | AK004569.1 | - | 11 - 12 | 1240 | 85 | |
| AK136292.1 | - | 3 - 4 | 1193 | 85 | ||
| BC147503.1 | - | 11 - 12 | 1273 | 9056386 | 85 | |
| BC158078.1 | - | 11 - 12 | 1273 | 9088170 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc40
Genomic Map