Assay Details
| Gene Symbol: Cdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027879.2 | NP_082155.1 | 8 - 9 | 1010 | 59 | |
| GenBank mRNA | AK004569.1 | - | 8 - 9 | 977 | 59 | |
| AK135411.1 | - | 8 - 9 | 977 | 59 | ||
| BC147503.1 | - | 8 - 9 | 1010 | 9056386 | 59 | |
| BC158078.1 | - | 8 - 9 | 1010 | 9088170 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc40
Genomic Map