Assay Details
| Gene Symbol: Grid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008167.2 | NP_032193.1 | 14 - 15 | 2367 | 65 | |
| GenBank mRNA | AK032669.1 | - | 2 - 3 | 191 | 65 | |
| BC139823.1 | - | 14 - 15 | 2371 | 40141838 | 65 | |
| D13266.1 | - | 14 - 15 | 2367 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grid2
Genomic Map