Assay Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077101.1 | - | 1319 | 105 | ||
| BC026926.1 | - | 6 - 7 | 1563 | 3586097 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

C87436
Genomic Map