Assay Details
| Gene Symbol: Slc6a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133661.3 | NP_598422.2 | 1 - 2 | 303 | 109 | |
| GenBank mRNA | BC019211.1 | - | 1 - 2 | 307 | 5053116 | 109 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019211.1, NM_133661.3 transcripts.
Set Membership: |
|

Slc6a12
Genomic Map