Assay Details
| Gene Symbol: Slc6a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009320.4 | NP_033346.2 | 1 - 2 | 249 | 72 | |
| GenBank mRNA | AK041141.1 | - | 1 - 2 | 222 | 72 | |
| AK143510.1 | - | 1 - 2 | 213 | 72 | ||
| AK162776.1 | - | 1 - 2 | 213 | 72 | ||
| BB636410.1 | - | 1 - 2 | 222 | 72 | ||
| BC015245.1 | - | 1 - 2 | 186 | 4242193 | 72 | |
| L03292.2 | - | 1 - 2 | 221 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041141.1, AK143510.1, BC015245.1, L03292.2, NM_009320.4 transcripts.
Set Membership: |
|

Slc6a6
Genomic Map