Assay Details
| Gene Symbol: Lrrc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015011.1 | - | 1 - 2 | 68 | 88 | |
| BC049601.1 | - | 1 - 2 | 75 | 6743951 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC049601.1 transcripts.
Set Membership: |
|

Lrrc23
Genomic Map