Assay Details
| Gene Symbol: Zbtb24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277229.1 | NP_001264158.1 | 3 - 4 | 1327 | 93 | |
| NM_153398.3 | NP_700447.2 | 3 - 4 | 1327 | 93 | ||
| GenBank mRNA | AK054097.1 | - | 3 - 4 | 1283 | 93 | |
| AK146986.1 | - | 3 - 4 | 1303 | 93 | ||
| AY210402.1 | - | 3 - 4 | 1301 | 93 | ||
| BC050933.1 | - | 3 - 4 | 1289 | 6811382 | 93 | |
| BC055367.1 | - | 3 - 4 | 1270 | 30056072 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb24
Genomic Map