Assay Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028099.4 | NP_082375.4 | 2 - 3 | 301 | 70 | |
| GenBank mRNA | AK040232.1 | - | 2 - 3 | 279 | 70 | |
| AK081629.1 | - | 2 - 3 | 278 | 70 | ||
| AK088941.1 | - | 2 - 3 | 298 | 70 | ||
| BC067028.1 | - | 2 - 3 | 255 | 30364073 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp11
Genomic Map