Assay Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028099.4 | NP_082375.4 | 3 - 4 | 443 | 193 | |
| GenBank mRNA | AK040232.1 | - | 3 - 4 | 421 | 193 | |
| AK081629.1 | - | 3 - 4 | 420 | 193 | ||
| AK088941.1 | - | 3 - 4 | 440 | 193 | ||
| BC067028.1 | - | 3 - 4 | 397 | 30364073 | 193 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp11
Genomic Map