Assay Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028099.4 | NP_082375.4 | 4 - 5 | 562 | 120 | |
| GenBank mRNA | AK081629.1 | - | 4 - 5 | 539 | 120 | |
| BC067028.1 | - | 4 - 5 | 516 | 30364073 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp11
Genomic Map