Assay Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028099.4 | NP_082375.4 | 5 - 6 | 616 | 71 | |
| GenBank mRNA | AK081629.1 | - | 5 - 6 | 593 | 71 | |
| BC028640.1 | - | 1 - 2 | 43 | 71 | ||
| BC067028.1 | - | 5 - 6 | 570 | 30364073 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC028640.1 transcripts.
Set Membership: |
|

Dusp11
Genomic Map